Learn More
Bachem 6-Azido-hexanoic acid
SDP
Catalog No. 501947397
Encompass_Preferred
6-Azido-hexanoic acid, 79598-53-1, C6H11N3O2, Mr 157.17, 5g. e-Azidocaproic acid. Building block for Staudinger ligation as well as for "click chemistry". This highly selective reaction can be applied for the conjugation of peptides with various molecules and for the cyclization of peptides. The azido moiety may also be considered as a protected amino group which is deblocked under reductive conditions.
Catalog No. 50-194-7397 Supplier Bachem Supplier No. 4066756.0005
Add to Cart
Add to Cart
6-Azido-hexanoic acid, 79598-53-1, C6H11N3O2, Mr 157.17, 5g. e-Azidocaproic acid. Building block for Staudinger ligation as well as for "click chemistry". This highly selective reaction can be applied for the conjugation of peptides with various molecules and for the cyclization of peptides. The azido moiety may also be considered as a protected amino group which is deblocked under reductive conditions.