Learn More
Bachem Fmoc-D-allo-Thr-OH
SDP
Supplier:  Bachem 4027554.0005
Encompass_Preferred
Fmoc-D-allo-Thr-OH, 130674-54-3, C19H19NO5, Mr 341.36, 5g. Building block for the SPPS of kahalalide F. The hydroxyl group of D-aThr is involved in the ester bond of this cyclic depsipeptide.
Catalog No. 50-194-7332