Learn More
Bachem Fmoc-Asp(EDANS)-OH
SDP
Catalog No. 501947123
Encompass_Preferred
Fmoc-Asp(EDANS)-OH, 182253-73-2, C31H29N3O8S, Mr 603.65, 0.5g. Building block for the synthesis of internally quenched fluorogenic peptide substrates. The fluorescent group EDANS can be efficiently quenched by the nonfluorescent DABCYL chromophore.
Catalog No. 50-194-7123 Supplier Bachem Supplier No. 4049431.0500
Fmoc-Asp(EDANS)-OH, 182253-73-2, C31H29N3O8S, Mr 603.65, 0.5g. Building block for the synthesis of internally quenched fluorogenic peptide substrates. The fluorescent group EDANS can be efficiently quenched by the nonfluorescent DABCYL chromophore.