Learn More
Bulbman Inc FHE/ESB 6V 20W G4 HALOGEN BULB
SDP
Catalog No. NC9023394
Encompass_Preferred
FHE Light Bulb, 6V, 20w, 0.375 in. dia., Base: G4 base, Overall Length: 1.25 in., Life: 2000 hr
Catalog No. NC9023394 Supplier Bulbman Inc Supplier No. 130813
FHE Light Bulb, 6V, 20w, 0.375 in. dia., Base: G4 base, Overall Length: 1.25 in., Life: 2000 hr