Learn More
Selleck Chemical LLC Bulevirtide Myrcludex B-5mg
SDP
Catalog No. 502389712
Encompass_Preferred
Bulevirtide (Myrcludex B) is a sodium-taurocholate co-transporting polypeptide (NTCP) receptor inhibitor with IC50 of ∼80 pM and inactivates NTCP function at concentrations far below those required to block bile salt transport.
Catalog No. 50-238-9712 Supplier Selleck Chemical LLC Supplier No. P11055MG
Bulevirtide (Myrcludex B) is a sodium-taurocholate co-transporting polypeptide (NTCP) receptor inhibitor with IC50 of ∼80 pM and inactivates NTCP function at concentrations far below those required to block bile salt transport.