Learn More
Poltex Bulk 3-Ply/N95 Mask Holder - Cam Lock
SDP
Catalog No. 01259135
Change view
Click to view available options
Mounting:
Tape Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-135 Tape Mount
01-259-153 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
N/A
Catalog No. 01-259-135 Supplier Poltex Supplier No. MASKBULKT
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A
Holds and dispenses earloop masks or cone masks including N95. Features a keyed camlock lid and adjustable internal dividers with Wall, Tape, or Magnet Mount options.
  • Holds and dispenses either earloop masks or cone masks (N95 or standard).
  • Comes with a keyed camlock to secure the lid, all keys are keyed the same.
  • Internal dividers slide in two ways to adjust to cone or earloop masks.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.
Specifications
Product Type Face Mask Dispenser
Dimensions (L x W x H) 5.6 x 8.5 x 9.8 in.
Includes High Bond Tape for Mounting
Packaging Quantity 1
Mounting Tape Mount
Holds Face Masks (3-ply/n95)
No. of Compartments 1