Learn More
Grainger Vestil Forklift Or Hoist Bulk Bag Lifter, Steel, 4000 Lb. Capacity, 1 ea.
Catalog No. 502774186
Encompass_Preferred
This forklift bulk bag lifter provides a reliable solution for lifting, transporting, and positioning FIBC bulk bags in warehouse, production, and material handling environments. Engineered for use with both forklifts and overhead hoists, the unit improves control and stability during the handling of heavy bulk loads such as powders, pellets, and sand. It features a rugged welded steel construction finished with a durable powder coat for long-term industrial service.
  • Capacity of 4,000 lb.
  • Integrated fork pockets and included restraint strap for secure attachment to lift trucks
  • Centrally located lifting hook for overhead hoist applications
  • Multiple lifting points for balanced load handling
  • Underside bag liner hook to assist with positioning
Catalog No. 50-277-4186 Supplier Grainger Supplier No. 172L64
This forklift bulk bag lifter provides a reliable solution for lifting, transporting, and positioning FIBC bulk bags in warehouse, production, and material handling environments. Engineered for use with both forklifts and overhead hoists, the unit improves control and stability during the handling of heavy bulk loads such as powders, pellets, and sand. It features a rugged welded steel construction finished with a durable powder coat for long-term industrial service.
  • Capacity of 4,000 lb.
  • Integrated fork pockets and included restraint strap for secure attachment to lift trucks
  • Centrally located lifting hook for overhead hoist applications
  • Multiple lifting points for balanced load handling
  • Underside bag liner hook to assist with positioning