Learn More
Alkali Scientific ASI™ Bulk Beads, Silica (glass), 0.1mm, acid washed, 200g
Supplier: Alkali Scientific RS71310
Bulk Beads, Silica (glass), 0.1mm, acid washed, 200g Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.