Learn More
Alkali Scientific ASI™ Bulk Beads, Silica (glass), 0.5mm, acid washed, 200g
SDP
Catalog No. 503027381
Encompass_Preferred
Bulk Beads, Silica (glass), 0.5mm, acid washed, 200g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-7381 Supplier Alkali Scientific Supplier No. RS71315
Bulk Beads, Silica (glass), 0.5mm, acid washed, 200g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.