Learn More
Alkali Scientific ASI™ Bulk Beads, Silica (glass), 1.0mm acid washed, 200g
SDP
Catalog No. 503028923
Encompass_Preferred
Bulk Beads, Silica (glass), 1.0mm acid washed, 200g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-8923 Supplier Alkali Scientific Supplier No. RS71311
Bulk Beads, Silica (glass), 1.0mm acid washed, 200g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.