Learn More
Alkali Scientific ASI™ Bulk Beads, Zirconium, 0.1mm, Triple-Pure Molecular Biology Grade, 250g
SDP
Catalog No. 503028452
Encompass_Preferred
Bulk Beads, Zirconium, 0.1mm, Triple-Pure Molecular Biology Grade, 250gCompatible with BeadBug™ Microtube   homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-8452 Supplier Alkali Scientific Supplier No. RS71311TP
Bulk Beads, Zirconium, 0.1mm, Triple-Pure Molecular Biology Grade, 250gCompatible with BeadBug™ Microtube   homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.