Learn More
Alkali Scientific ASI™ Bulk Beads, Zirconium, 0.5mm, Triple-Pure Molecular Biology Grade, 250g
SDP
Catalog No. 503027628
Encompass_Preferred
Bulk Beads, Zirconium, 0.5mm, Triple-Pure Molecular Biology Grade, 250g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-7628 Supplier Alkali Scientific Supplier No. RS71315TP
Bulk Beads, Zirconium, 0.5mm, Triple-Pure Molecular Biology Grade, 250g   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.