Learn More
Alkali Scientific ASI™ Bulk Beads, Zirconium, 1.0mm, Triple-Pure Molecular Biology Grade, 250g
SDP
Catalog No. 503026910
Encompass_Preferred
Bulk Beads, Zirconium, 1.0mm, Triple-Pure Molecular Biology Grade, 250g Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.  
Catalog No. 50-302-6910 Supplier Alkali Scientific Supplier No. RS71312TP
Bulk Beads, Zirconium, 1.0mm, Triple-Pure Molecular Biology Grade, 250g Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.  