Learn More
SAI Infusion Technologies 30 Gauge, 1.5" Blunt Needles - Non-sterile, 1000/pk
SDP
Catalog No. NC3718861
Encompass_Preferred
30G1.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC3718861 Supplier SAI Infusion Technologies Supplier No. B30150 1000 BULK
30G1.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.