Learn More
Poltex Bulk Earloop Mask Disp Mount
SDP
Catalog No. 22000523
Change view
Click to view available options
Includes:
Powerful Magnets for Mounting
Screws and Anchors for Mounting
VHB Tape for Mounting
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-523 VHB Tape for Mounting
22-000-521 Powerful Magnets for Mounting
22-000-522 Screws and Anchors for Mounting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-523 Supplier Poltex Supplier No. MSKFEEDT
  • $77.20
    $64.65 / Each of 1
    Save $12.55
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Holds a stack of bulk earloop masks. Slanted internal ramp helps to feed masks toward the front. Durable, cleanable PETG construction. Choose from VHB tape mount (very high bond tape included), or magnet mount (powerful magnets included).
Holds a stack of bulk earloop masks. Slanted internal ramp helps to feed masks toward the front. Choose from magnet mount, VHB tape mount, or wall mount.
  • Durable, cleanable PETG construction
  • VHB tape mount (very high bond tape included)
  • Wall mount (screws and anchors included)
  • Magnet mount (powerful magnets included)
Specifications
Product Type Face Mask Dispenser
Includes VHB Tape for Mounting