Learn More
Andwin Scientific BULK LTS 10ML TIP 200/PK
SDP
Catalog No. NC0266091
Encompass_Preferred
The BioClean tips have a volume capacity of 10 mL. All tips are free of DNA, DNase, RNase, pyrogen, ATP, and PCR inhibitors. This product is sold in bulk quantity of 200 tips per pack. When purchased in bulk, the tips are packaged in a resealable zip-lock bags.
Catalog No. NC0266091 Supplier Andwin Scientific Supplier No. RCL10MLQ
The BioClean tips have a volume capacity of 10 mL. All tips are free of DNA, DNase, RNase, pyrogen, ATP, and PCR inhibitors. This product is sold in bulk quantity of 200 tips per pack. When purchased in bulk, the tips are packaged in a resealable zip-lock bags.