Learn More
Alkali Scientific ASI™ Bulk Pipet Tips, 10uL Extra Long
SDP
Catalog No. 503026521
Encompass_Preferred
Bulk Pipet Tips by ASI are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipet Tip line is packaged Rnase/Dnase, pyrogen free and available in both sterile and non-sterile bags of 1000 tips. Choose the right size for your research: 10µl, 100µl, 200µl, 300µl or 1000µl volumes. Fine Point Design Certified Rnase, Dnase & Pyrogen Free Universal Fit Ultra Clear Polypropylene Withstands Organic Solvents Low-Retention 1000/Pack
Catalog No. 50-302-6521 Supplier Alkali Scientific Supplier No. BT1010
Bulk Pipet Tips by ASI are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipet Tip line is packaged Rnase/Dnase, pyrogen free and available in both sterile and non-sterile bags of 1000 tips. Choose the right size for your research: 10µl, 100µl, 200µl, 300µl or 1000µl volumes. Fine Point Design Certified Rnase, Dnase & Pyrogen Free Universal Fit Ultra Clear Polypropylene Withstands Organic Solvents Low-Retention 1000/Pack