Learn More
Bulbman Inc Bulk Quantity, 10 Osram 54842 EKE, 150 watt
SDP
Catalog No. 501253636
Encompass_Preferred
Bulk Qty. 10 Osram 54842 EKE: EKE is a halogen specialty light bulb. The EKE specification include: 21 volt 150 watt 200 hour rated life CC6 filament vertical (plus or minus 45 degrees) base down operating position MR16D quartz glass dichroic reflector and GX5.3 bi-pin base. The EKE is used in medical / scientific microscopes photo printers and projectors. The EKE is a possible substitute for ANSI ENB. Possible substitute: EJV (25 hour life more heat).
Catalog No. 50-125-3636 Supplier Bulbman Inc Supplier No. 10340810
Bulk Qty. 10 Osram 54842 EKE: EKE is a halogen specialty light bulb. The EKE specification include: 21 volt 150 watt 200 hour rated life CC6 filament vertical (plus or minus 45 degrees) base down operating position MR16D quartz glass dichroic reflector and GX5.3 bi-pin base. The EKE is used in medical / scientific microscopes photo printers and projectors. The EKE is a possible substitute for ANSI ENB. Possible substitute: EJV (25 hour life more heat).