Learn More
Bulbman Inc Bulk Quantity, 10 Osram 64275, 35 watt
SDP
Catalog No. 501253643
Encompass_Preferred
Bulk Qty. 10 Osram 64275: This Osram 64275, 54258 bulb uses 35 watts and 6 volts. It has a bipin base and an average rated life of 50 hours. It has a T3 lamp shape and is a halogen bulb
Catalog No. 50-125-3643 Supplier Bulbman Inc Supplier No. 11738610
Bulk Qty. 10 Osram 64275: This Osram 64275, 54258 bulb uses 35 watts and 6 volts. It has a bipin base and an average rated life of 50 hours. It has a T3 lamp shape and is a halogen bulb