Learn More
Bulbman Inc BULK QTY. 10 USHIO EKZ
SDP
Catalog No. 501253637
Encompass_Preferred
Bulk Qty. 10 Ushio EKZ: Tungsten Halogen, 50mm Diameter Dichroic Coated Reflector. This Ushio EKZ Lamp 30W/10.8V is a tungsten halogen bulb with an MR-16 reflector which is compatible with fixtures that have a GX5.3 base and at least 30 watt rating. It has a color temperature of 3100K and an average life of 200 hours.
Catalog No. 50-125-3637 Supplier Bulbman Inc Supplier No. 11096310
Bulk Qty. 10 Ushio EKZ: Tungsten Halogen, 50mm Diameter Dichroic Coated Reflector. This Ushio EKZ Lamp 30W/10.8V is a tungsten halogen bulb with an MR-16 reflector which is compatible with fixtures that have a GX5.3 base and at least 30 watt rating. It has a color temperature of 3100K and an average life of 200 hours.