Learn More
Bulbman Inc BULK QTY. 5 HBO100W/2 69217
SDP
Catalog No. 501253644
Encompass_Preferred
Bulk Qty. 5 HBO100W/2: Osram 69217 HBO100W/2 mercury short arc lamp is used in microscope UV light curing and photo-polymerization applications.
Catalog No. 50-125-3644 Supplier Bulbman Inc Supplier No. 1051715
Bulk Qty. 5 HBO100W/2: Osram 69217 HBO100W/2 mercury short arc lamp is used in microscope UV light curing and photo-polymerization applications.