Learn More
Biosure Inc BULK SHEATH SOLUTION 2X10L/CS
Catalog No. NC9801719
Encompass
BioSure Bulk 8X Sheath Solution is a buffered, azide-free reagent. The product contains an antibacterial-antifungal preservative (not azide) and may not be suitable for cells that are cultured following sorting.
Catalog No. NC9801719 Supplier Biosure Inc Supplier No. 1023
BioSure Bulk 8X Sheath Solution is a buffered, azide-free reagent. The product contains an antibacterial-antifungal preservative (not azide) and may not be suitable for cells that are cultured following sorting.