Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl Bulk Shelving Bottom Cross Bar

Catalog No. 11222158
Change view
Click to view available options
Width (English):
36 in.
48 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-222-158 36 in.
11-222-159 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-222-158 Supplier Wanzl Supplier No. T.BCRB36CB
Add to Cart
Add to Cart

Bottom Cross Bar for use with bulk shelving.

  • Bottom Cross Bar
  • Replacement part for bulk shelving system

Specifications

Color Chrome
Includes 1 Bottom Cross Bar
For Use With (Application) Shelving
Width (English) 36 in.
Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.