Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Bulk Storage Bin - Front Drop Door
SDP

Catalog No. 01259202
Change view
Click to view available options
Mounting Type:
Magnet
Tape
Wall
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
01-259-202 Tape
01-259-165 Magnet
01-259-166 Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-202 Supplier Poltex Supplier No. BINDISPT
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

Bulk storage bin for holding lab or clinic consumables such as microtubes, conical tubes, caps, and tips. Fill from the top and dispense from the front, with multiple mounting options for flexible placement.

  • Benchtop storage bin for bulk lab or clinic consumables
  • Ideal for microtubes, conical tubes, caps, pipette tips, and more
  • Top-fill design for easy loading
  • Front dispensing opening for convenient access
  • Available in Wall Mount (screws and anchors included)
  • Available in HB Tape Mount (high-bond tape included)
  • Available in Magnet Mount (powerful magnets included)
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Product Type Organizer
Mounting Type Tape
Holds Misc Bulk Lab Items, Microtubes, Conical Tubes, Caps, Tips
Color Clear
Quantity 1 Ea.
Includes High Bond Tape
No. of Shelves None
No. of Doors 2
Dimensions (L x W x H) 6.8 x 7 x 7.1 in.
Autoclavable Not Autoclavable
For Use With (Equipment) Microtubes, Conical Tubes, Caps, Tips
Material PETG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.