Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bead Mill Homogenizer Accessory, Tubes with Screw Caps and O-Rings
GREENER_CHOICE

Catalog No. 15340162 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Non-reinforced Construction
Reinforced Construction
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15-340-162 Reinforced Construction
15-340-161 Non-reinforced Construction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-340-162
  • PRODUCT PROMOTION
    $665.00
    $337.09 / Pack of 500

    Save $327.91

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.

  • RNase/DNase free
  • Processing range: 300μL to 1.8mL
  • Beads not included

Specifications

Capacity (Metric) 2 mL
Autoclavable Autoclavable
Color White/Opaque
Dnase Free Yes
For Use With (Equipment) Bead Mill 4 and 24 Homogenizers
Includes Reinforced Construction
Packaging Quantity 500/Pk.
RNase Free Yes
Product Type Bead Mill Homogenization Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.