Learn More
Waters Corp Waters Elbow, Male, Bulkhead Fitting, 4 mm x 12
Catalog No. 50466580 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This fitting is an elbow, male bulkhead type, precisely specified as 4 mm x 12. It is designed to facilitate connections through panels and is suitable for various applications.
  • Facilitates directional changes in fluid paths.
  • Features a male connection for secure fitting.
  • Designed with a bulkhead configuration for panel-through installations.
  • Manufactured with dimensions of 4 mm x 12 for specific system requirements.
Catalog No. 50-466-580 Supplier Waters Corp Supplier No. 700003044
This fitting is an elbow, male bulkhead type, precisely specified as 4 mm x 12. It is designed to facilitate connections through panels and is suitable for various applications.
  • Facilitates directional changes in fluid paths.
  • Features a male connection for secure fitting.
  • Designed with a bulkhead configuration for panel-through installations.
  • Manufactured with dimensions of 4 mm x 12 for specific system requirements.