Learn More
Bullard™ 3000R Series Safety Cap
SDP
Catalog No. 19086337 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Red
Tan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-086337 Tan
19-052684 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-086337 Supplier Bullard™ Supplier No. 30TNR
Add to Cart
Add to Cart
Classic cap-style hard hat
  • Six-point adjustable Flex-Gear™ suspension with seamless 1 in. nylon crown straps
  • Vertical height adjustment provides custom fit
  • Replaceable absorbent cotton brow pad, a nape cushion, and a large, easily gripable sizing knob
  • Accessory slots accept most attachments
Compliance
Meets ANSI Z89.1-2003, Type 1, Class E and G
Specifications
Color Tan
Quantity 1 Ea.
Suspension Type 6-Point Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.