Learn More
Bullard™ SAR System, 41 Series Portable Filtration and CO Monitoring Bundle
SDP
Catalog No. 19824415 Shop All Bullard Company Products
Change view
Click to view available options
Bundle includes CC20 Respirator
Catalog No.
19-824-415 Bundle includes CC20 Respirator
1 options
This item is not returnable. View return policy
Catalog No. 19-824-415 Supplier Bullard™ Supplier No. COHP41P2CC20
This item is not returnable. View return policy
Portable filtration bundle includes CC20 Respirator (not shown).
Specifications
Includes CC20 respirator, heavy duty 41 Series filter with 7-stage filtration, CO monitor with auto calibration, audible and visual alarms for high CO and low battery, and 6-foot 110 VAC power cord
Model COHP41P2CC20