Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Accessories and Replacement Parts for Hard Hat
SDP

Catalog No. 19150215 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
Hard Hats
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-150-215 Hard Hats
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-215 Supplier Bullard™ Supplier No. CP2
  • $13.75
    $8.65 / Each

    Save $5.10

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $118.00
    $75.65 / Case of 10 EA

    Save $42.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with head and face protection

Clear polycarbonate replacement chin protector.

  • Material: Polycarbonate
  • Color: Clear
  • Compatibility: For use with Bullard 840PX2, 840PX2W3, 840PX2W5, 840MX2 and 840TX2 models

Specifications

Color Clear
For Use With (Equipment) Hard Hats
Material Polycarbonate
Product Type Chin protector

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.