Learn More
Bullard™ Accessories for Spectrum Series Airline Respirator System: Respirator Assemblies
SDP
Catalog No. 19824323 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Assembly for Spectrum-PD
Facepiece for Spectrum-PD
Facepiece for Spectrum-PDE10
Facepiece for Spectrum-PDE5
Includes:
Medium/large facepiece, pdr45 pressure demand regulator, bha belt harness assembly, ca30 coupler assembly
Medium/large facepiece, regulator including bypass and 5 min. escape assembly with ca30 coupler assembly
Small facepiece, PDR45 pressure demand regulator, BHA belt harness assembly, CA30 coupler assembly
Small facepiece, RA45B regulator assembly (including 1st and 2nd stage regulators with bypass), harness assembly (including 10 min. ESCBA cylinder), CA30 coupler assembly
Small facepiece, regulator including bypass and 5 min. escape assembly with CA30 coupler assembly
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-824-323 Facepiece for Spectrum-PD Small facepiece, PDR45 pressure demand regulator, BHA belt harness assembly, CA30 coupler assembly
19-824-321 Facepiece for Spectrum-PDE5 Small facepiece, regulator including bypass and 5 min. escape assembly with CA30 coupler assembly
19-824-320 Facepiece for Spectrum-PDE5 Medium/large facepiece, regulator including bypass and 5 min. escape assembly with ca30 coupler assembly
19-824-319 Facepiece for Spectrum-PDE10 Small facepiece, RA45B regulator assembly (including 1st and 2nd stage regulators with bypass), harness assembly (including 10 min. ESCBA cylinder), CA30 coupler assembly
19-824-322 Assembly for Spectrum-PD Medium/large facepiece, pdr45 pressure demand regulator, bha belt harness assembly, ca30 coupler assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-824-323 Supplier Bullard™ Supplier No. SPECPDSA
Used for respiratory protection
Specifications
Product Type Facepiece for Spectrum-PD
Size Small
For Use With (Equipment) Spectrum series airline respirator system
Includes Small facepiece, PDR45 pressure demand regulator, BHA belt harness assembly, CA30 coupler assembly
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.