Learn More
Bullard™ High-Pressure Full Face Assembly for Spectrum Series
SDP
Catalog No. 19807908 Shop All Bullard Company Products
Change view
Click to view available options
Small continuous flow
Catalog No.
19-807-908 Small continuous flow
1 options
Catalog No. 19-807-908 Supplier Bullard™ Supplier No. SPEC40S
High-pressure continuous flow mask size small. For use with Spectrum series respriators.
Specifications
For Use With (Equipment) Spectrum Series Airline Respirator System
Includes Small Facepiece, Breathing Tube Assembly with 0.5 in. Industrial Interchange (Hansen compatible) Nipple, Nylon Belt
Model SPEC40S
Size Small
Product Type Continuous Flow Facepiece