Learn More
MSC Bullard Acetate Face Shield, 8 x 0.04 in, 1 ea
Catalog No. 5000782627
Encompass
This multi-fit acetate face shield visor provides protection against chemical splashes and general impact. It is designed for use with compatible face shield brackets, offering clear vision and durability for various applications.
  • Clear acetate construction
  • Resists chemical splashes
  • Meets ANSI Z87.1 standards
  • Designed for multi-fit face shield brackets
  • Provides 8 in height and 0.04 in thickness
Catalog No. 50-007-82627 Supplier MSC Supplier No. 85404390
This multi-fit acetate face shield visor provides protection against chemical splashes and general impact. It is designed for use with compatible face shield brackets, offering clear vision and durability for various applications.
  • Clear acetate construction
  • Resists chemical splashes
  • Meets ANSI Z87.1 standards
  • Designed for multi-fit face shield brackets
  • Provides 8 in height and 0.04 in thickness