Learn More
Bullard™ Advent™ EMS Safety Helmet
SDP
Catalog No. 17358C Shop All Bullard Company Products
Change view
Click to view available options
Red
Catalog No.
17-358C Red
1 options
This item is not returnable. View return policy
Catalog No. 17-358C Supplier Bullard™ Supplier No. A1RDS/DEL
This item is not returnable. View return policy
Helmet of choice for urban, industrial, and confined-space rescue operations
Used by FEMA and USAR teams. Ideal for paramedics, emergency medical technicians, industrial hazard response teams, high-angle rescues, and wilderness operations.
  • Polycarbonate outer shell and inner foam liner for superior impact protection for the top, front, back, and side of the head
  • Easy to adjust and wear with comfortable crown pad and two-point chin strap
  • Flex-Gear™ ratchet suspension system fits sizes 6.5 to 8
Order Info
Weighs only 27 oz. Custom printing of logos or reflective strips also available by special order.
Specifications
Color Red
Material Vinyl
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.