Learn More
Bullard™ Advent™ Series Firefighter Helmets
SDP
Catalog No. 19824460 Shop All Bullard Company Products
Change view
Click to view available options
Suspension Type:
8-pt. Flex Gear ratchet suspension
Catalog No. Suspension Type
19-824-460 8-pt. Flex Gear ratchet suspension
1 options
Catalog No. 19-824-460 Supplier Bullard™ Supplier No. A2WHS
  • $305.00
    $241.65 / Each of 1
    Save $63.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Specifications
Certifications/Compliance Meets/exceeds ANSI Z89.1-2003 Type II, Class E and G requirements
Construction Outer shell: polycarbonate alloy; EPS foam impact liner and crown cushion
Color White
Liner Material EPS foam
Suspension Type 8-pt. Flex Gear ratchet suspension