Learn More
Bullard™ Assemblies and Accessories for CC20 Series Supplied Air Respirator
SDP
Catalog No. 19824394 Shop All Bullard Company Products
Change view
Click to view available options
Adjustable flow control valve; Vinyl decon belt
Catalog No.
19-824-394 Adjustable flow control valve; Vinyl decon belt
1 options
Catalog No. 19-824-394 Supplier Bullard™ Supplier No. F40VDB
Used for respiratory protection
Specifications
For Use With (Equipment) CC20 Series
Material Steel
Includes 0.25 in. Industrial Interchange (Hansen Compatible) Quick-disconnect Nipple, Threaded Hose Connector, Vinyl Decon Belt
Model F40VDB
Product Type Adjustable Flow Control Valve