Learn More
Bullard™ Breathing Air Supply Hose Nipple
SDP
Catalog No. 19824358 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
0.25 in. Bayonet
0.25 in. Schrader
Includes:
Female pipe thread
V12 hose adapter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type Includes
19-824-358 0.25 in. Schrader Female pipe thread
19-824-355 0.25 in. Bayonet V12 hose adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-358 Supplier Bullard™ Supplier No. S19433
Quick-disconnect nipples are for use with V10 airline hoses.
  • Accessory for use with V10 hoses only
  • 0.25 in. nipple
  • Available in bayonet or schrader steel
Specifications
Connection Type 0.25 in. Schrader
Material Schrader
Product Type Hose Nipple
For Use With (Equipment) V10 hoses
Includes Female pipe thread
Model S19433
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.