Learn More
MSC BULLARD C33 Hard Hat, Full Brim, Ratchet Suspension, 1 ea
Catalog No. 5000794615
Encompass
The 3-Rib C33 is a full brim hard hat featuring a 6-point Flex-Gear® ratchet suspension, engineered to protect against falling objects and impacts. Its wide profile offers all-around protection from sun, rain, and dust, ideal for demanding job sites. This hard hat meets ANSI/ISEA Z89.1 Type I Class E and G, and CSA Z94.1 standards.
  • Protects against falling objects and impacts
  • Equipped with 6-point Flex-Gear® ratchet suspension for secure fit
  • ANSI/ISEA Z89.1 Type I, Class E and G certified
  • CSA Z94.1 Type 1, Class E and G certified
  • Lightweight with low profile and center of gravity
  • Adjustable for hat sizes 6.5 to 8 (21 to 25 in)
  • Vertical adjustment capability (two front, two rear)
  • Permanent under-brim glare guard
  • Two-year warranty
Catalog No. 50-007-94615 Supplier MSC Supplier No. 71275473
The 3-Rib C33 is a full brim hard hat featuring a 6-point Flex-Gear® ratchet suspension, engineered to protect against falling objects and impacts. Its wide profile offers all-around protection from sun, rain, and dust, ideal for demanding job sites. This hard hat meets ANSI/ISEA Z89.1 Type I Class E and G, and CSA Z94.1 standards.
  • Protects against falling objects and impacts
  • Equipped with 6-point Flex-Gear® ratchet suspension for secure fit
  • ANSI/ISEA Z89.1 Type I, Class E and G certified
  • CSA Z94.1 Type 1, Class E and G certified
  • Lightweight with low profile and center of gravity
  • Adjustable for hat sizes 6.5 to 8 (21 to 25 in)
  • Vertical adjustment capability (two front, two rear)
  • Permanent under-brim glare guard
  • Two-year warranty