Learn More
Bullard™ Assemblies for DC5040 Series cooling vests and quick-disconnect nipples.
SDP
Catalog No. 19400017 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
Bayonet
CEJN
Schrader
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
19-400-017 CEJN
19-400-013 Schrader
19-400-018 Bayonet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-400-017 Supplier Bullard™ Supplier No. DC5047
Add to Cart
Add to Cart
Dual-Cool adjustable cooling tubes are for use with GenVX, 88VX, CC20, and GR50 series airline respirators.
  • Supplied-Air Respirator (SAR) body temperature management accessory
  • Dual-Cool tubes DC50
  • DC5041 - 0.25 in. schrader QD nipple
  • DC5047 - 0.25 in. CEJN QD nipple
  • DC5048 - 0.25 in. Bayonet QD Nipple
Specifications
Certifications/Compliance CE, NIOSH
Connection Type CEJN
Material CEJN
Product Type Hose Nipple
For Use With (Equipment) Dual-cool vests, 88VX, CC20, GR50 series airline respirators
Includes Heat shield
Model DC5047
Diameter (English) Interior 0.25 in.
Diameter (Metric) Interior 0.63 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.