Learn More
Bullard™ Dual-Cool™ Climate Control Assemblies, Quick-Disconnect Nipples
SDP
Catalog No. 19822174 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Industrial Interchange Steel
Snap-tite Brass
Snap-tite Stainless Steel
Snap-tite Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-822-174 Industrial Interchange Steel
19-400-014 Snap-tite Steel
19-400-015 Snap-tite Brass
19-400-016 Snap-tite Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-822-174 Supplier Bullard™ Supplier No. DC5040
Add to Cart
Add to Cart
Climate control device with quick-disconnect nipple in a variety of coupling type materials.
Specifications
Certifications/Compliance CE, NIOSH
For Use With (Equipment) DC Vests of GenVX, 88VX, CC20 and GR50 Series Airline Respirator
Material Industrial Interchange Steel
Includes CH60 Connector Hose and Nylon Belt
Model DC5040
Product Type Climate Control Device
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.