Learn More
Bullard™ EVA PAPR System with Double Bib Nomex Grinding Hoods
SDP
Catalog No. 17989617 Shop All Bullard Company Products
Catalog No. 17-989-617 Supplier Bullard™ Supplier No. EVAGRHT
Add to Cart
Add to Cart
Ergonomically designed PAPR is ideal for use in hazardous locations.
  • PAPR with double bib nomex grinding quick attach T-Series hood
  • PC lens and equipped with Active Flow Technology System
  • Offers 1,000 Assigned Protection Factor (APF)
  • NIOSH approved
  • Set includes blower, belt, battery, charger, breathing tube, air flow indicator, FR head top with ratchet suspension
Specifications
Model EVAGRHT
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.