Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Blower for EVA™ Powered Air Purifying Respirator
SDP

Catalog No. 17986275 Shop All Bullard Company Products
Change view
Click to view available options
Battery Life:
4 to 16 hours
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Life
17-986-275 4 to 16 hours
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-986-275 Supplier Bullard™ Supplier No. EVA1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constant flow blower is designed to complete the EVA powered air purifying respirator when assembled with a hood, breathing tube, and filter cartridge.

  • Blower assembly designed for EVA PAPR
  • Choice of assembly configurations 
  • NIOSH approved EVAHL powered air purifying system consists of four parts:
    • Hood
    • Breathing Tube
    • Blower assembly
    • Filter cartridge
  • User controlled, choose from high or low speed
  • Brushless motor technology

Specifications

Battery Life 4 to 16 hours
Battery Type Rechargeable Lithium Polymer
Certifications/Compliance NIOSH, ANSI/ISA 2.12.01-2007, CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
Connection Type Belt
Color Black, White
Product Type Motor Blower
Flow Rate 7.0 to 8.5 CFM
For Use With (Application) Pharmaceutical manufacturing, healthcare, laboratories, painting, powder coating, general industries
For Use With (Equipment) EVA PAPR Systems
Includes Blower only
Model EVA1
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.