Learn More
Bullard™ Exhalation Valve Replacement Part
SDP
Catalog No. 19824327 Shop All Bullard Company Products
Change view
Click to view available options
Replacement exhalation valve
Catalog No.
19-824-327 Replacement exhalation valve
1 options
This item is not returnable. View return policy
Catalog No. 19-824-327 Supplier Bullard™ Supplier No. 6059
This item is not returnable. View return policy
Replacement exhalation valve kit for Spectrum series facepiece.
  • Replacement kit
  • For use with Spectrum series respirators
  • Sold 5/Pk.
Specifications
Product Type Exhalation Valve
Model 6059
Quantity 5/Pk.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.