Learn More
Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
SDP
Catalog No. 18998002 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
Contoured-fit, blue twist lock connection
Narrow profile, black quick-attach twist lock connection
Narrow profile, blue twist lock connection
Narrow profile, green twist lock connection
Traditional fit, Quick-attach twist lock connection
Size:
Small
Medium
Large
X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
18-998-002 Narrow profile, black quick-attach twist lock connection Large
19-824-430 Narrow profile, green twist lock connection Small
17-918-681 Traditional fit, Quick-attach twist lock connection X-Large
18-998-003 Narrow profile, blue twist lock connection Medium
19-021-233B Traditional fit, Quick-attach twist lock connection Medium
19-021-233C Traditional fit, Quick-attach twist lock connection -
19-098-042 Contoured-fit, blue twist lock connection -
19-098-041 Contoured-fit, blue twist lock connection Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-998-002 Supplier Bullard™ Supplier No. 20LF2L
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Loose fitting facepieces provide comfortable and effective protection from a wide range of respiratory hazards.
  • Loose fitting facepiece
  • Head covers made of Tychem
  • Sewn-in headband suspension
  • Twist-lock breathing tube connection
  • For use with PAPR and SAR systems
  • Choose from a variety of sizes and color connections
  • Acetate lens material
  • NIOSH approved
Specifications
Assigned Protection Factor (APF) 25
Product Type Loose Fitting Facepiece
Size Large
For Use With (Application) Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare
For Use With (Equipment) Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator
Includes Narrow profile, black quick-attach twist lock connection
Model 20LF2L
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.