Learn More
Bullard™ Flow Control Assemblies for CC20 Series Airline Respirator
SDP
Catalog No. 19824385 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Brass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-824-385 Stainless Steel
19-824-384 Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-385 Supplier Bullard™ Supplier No. F30S
Add to Cart
Add to Cart
Constant flow fittings for CC20 Series airline respirators.
  • Airline respirator accessory
  • Constant flow fitting
  • For use with CC20
  • 0.25 in. industrial interchange
Specifications
Material Stainless Steel
Product Type Constant Flow Control Valve
For Use With (Equipment) CC20 Series Compressed Air Respirator
Includes Industrial interchange (Hansen-compatible) quick-disconnect nipple, nylon belt, threaded hose connector
Model F30S
Diameter (English) Interior 0.25 in.
Diameter (Metric) Interior 0.63 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.