Learn More
Bullard™ Constant Flow Assemblies for RT Series Airline Respirators
SDP
Catalog No. 19824388 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
0.25 in. Bayonet Nipple
0.25 in. CEJN Nipple
0.25 in. Industrial Interchange (Hansen-compatible) Nipple
0.25 in. Snap-Tite Stainless Steel Nipple
0.25 in. Snap-Tite Steel Nipple
0.5 in. Industrial Interchange Brass Nipple
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
19-824-388 0.25 in. Snap-Tite Steel Nipple
19-807-854 0.25 in. Industrial Interchange (Hansen-compatible) Nipple
19-824-389 0.25 in. Snap-Tite Stainless Steel Nipple
19-824-390 0.5 in. Industrial Interchange Brass Nipple
19-824-391 0.25 in. CEJN Nipple
19-824-392 0.25 in. Bayonet Nipple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-824-388 Supplier Bullard™ Supplier No. F32
Add to Cart
Edge
Add to Cart
Flow control assembly for use with RT Series respirator, does not include breathing tube.
  • Constant flow accessory
  • For use with RT series airline respirator
  • Flow control assemblies
  • Nipple types include bayonet, CEJN, industrial interchange, schrader, or snap-tite
  • Does not include breathing tube
Specifications
Connection Type 0.25 in. Snap-Tite Steel Nipple
Controller Type Constant Flow
Material Stainless Steel
Product Type Flow Control Assembly
For Use With (Equipment) RT Series Airline Respirator
Model F32
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.