Learn More
Bullard™ Flow Control Valve for Airline Respirator
SDP
Catalog No. 19824387 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
No belt
Vinyl belt
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-824-387 Vinyl belt
19-824-386 No belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-387 Supplier Bullard™ Supplier No. F30VDB
Specifications
Product Type Flow Control Valve
For Use With (Application) Compressed air capabilities
For Use With (Equipment) 88VXBT, L99BT, 20BT or 20LFBT
Includes Vinyl belt
Model F30VDB
Diameter (English) Interior 0.25 in.
Diameter (Metric) Interior 0.63 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.