Learn More
Bullard™ Hardhat and Cap Face Shield Aluminum Brackets
SDP
Catalog No. 19021320 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
C30 Series Cap-style Hard Hat Models, Faceshield
C30, S61, S62, S51, 911C Cap-style, 911H Full Brim Series and Bump Cap Type II Fard Hat, Faceshield
C33, C34, S71 Full Brim Hard Hat
Product Type:
Gold Line Bracket
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
19-021320 C30, S61, S62, S51, 911C Cap-style, 911H Full Brim Series and Bump Cap Type II Fard Hat, Faceshield -
17-360 C30 Series Cap-style Hard Hat Models, Faceshield -
19-021327 C33, C34, S71 Full Brim Hard Hat Gold Line Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-021320 Supplier Bullard™ Supplier No. 100G
This item is not returnable. View return policy
Rugged brackets are ideal for high-heat industrial applications
Aluminum Brackets: Gold Line™ brackets are constructed of rugged, gold anodized aluminum. Highly resistant to corrosion and heat. Ideal for chemical handling and high heat industrial applications. Band- or slot-mounted.
Specifications
For Use With (Equipment) C30, S61, S62, S51, 911C Cap-style, 911H Full Brim Series and Bump Cap Type II Fard Hat, Faceshield