Learn More
Bullard™ HC2400 Hot/Cool Climate Control Tube
SDP
Catalog No. 19400000 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
Flow Control Valve, Belt Bracket, Belt and Heat Shield
Catalog No. Includes
19-400-000 Flow Control Valve, Belt Bracket, Belt and Heat Shield
1 options
Catalog No. 19-400-000 Supplier Bullard™ Supplier No. HC2400
  • $774.00
    $547.65 / Each of 1
    Save $226.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
NIOSH-approved accessory for use with certain SAR systems to supply a continuous flow or warm or cool air.
  • Includes:
    • Hot/Cold Tube
    • Flow Control Valve
    • Belt Bracket
    • Belt and Heat Shield
Specifications
Certifications/Compliance NIOSH
For Use With (Equipment) Series CC20 Airline Respirator Hoods
Includes Flow Control Valve, Belt Bracket, Belt and Heat Shield
Model HC2400
Product Type Hot/Cold Tube