Learn More
Bullard™ Isotherm™ 2 Cooling Vest
SDP
Catalog No. 19033638B Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Isotherm Cooling Vest
Replacement Cool Pack
For Use With (Application):
For bullard isotherm 2 cooling vest
For practical use in the field
Quantity:
2 Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application) Quantity
19-033-638B Isotherm Cooling Vest For practical use in the field Each
19-033-638D Replacement Cool Pack For bullard isotherm 2 cooling vest 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-033-638B Supplier Bullard™ Supplier No. ISO2
  • $912.00
    $507.65 / Each of 1
    Save $404.35
    44% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Interchangeable front/back cooling system reduces heat-related illness
Adjustable vest comes with one set of cooling packs filled with nontoxic, noncarcinogenic cooling agent. The interchangeable front/back cooling system reduces heat-related illness without body shock from extreme temperature differential. Replacement cool packs available to reduce downtime.
  • Maintains constant temperature of 13°C (55°F)
  • Vest provides up to 2 1/2 hours of cooling power
  • Made from flame-retardant material
  • Cooling packs recharge in 20 minutes in a cooler of ice water without condensation
Order Info
  • Weight, 6 1/2 lb. (2.9kg)
Specifications
Product Type Isotherm Cooling Vest
Size Medium/Large
Certifications/Compliance ANSI/ISEA 107-2010 Class 2
Cooling Time 2.5 hours
For Use With (Application) For practical use in the field
Quantity Each