Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Mylar Lens Cover for PC90 Respirators
SDP

Catalog No. 18998000 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
PA20 Series
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-998-000 PA20 Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-998-000 Supplier Bullard™ Supplier No. 20LC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lens covers attaches to PAPR faceshield to protect from damage.

  • Fit over facepieces of hood-style respirators to protect from scratches, overspray and other damage
  • Disposable 25/pack
  • Adhesive edges

Specifications

Color Clear
Disposable Single-use
Material Tychem QC
Product Type Lens Cover
For Use With (Equipment) PA20 Series
Model 20 LC

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.